Anti-C4orf49 Rabbit Polyclonal Antibody

Supplier: Proteintech
22569-1-AP-150UL
10083-862EA 402 USD
10083-862
Anti-C4orf49 Rabbit Polyclonal Antibody
Antibodies
The protein MGARP, also named as C4ORF49, CESP1, HUMMR, OSAP and GS3582, plays a role in the trafficking of mitochondria along microtubules. It regulates the kinesin-mediated axonal transport of mitochondria to nerve terminals along microtubules during hypoxia. MGARP is localized to the mitochondria in cultured human CE, and its N-terminal seems to be processed after mitochondrial import. The MW of MGARP is about 33-36 kDa.

Western Blot:SKOV-3 Cells, 1:500-1:5000; IHC: Human ovary Tissue, 1:20-1:200

Type: Primary
Antigen: C4orf49
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR