Anti-CEP290 Rabbit Polyclonal Antibody

Supplier: Proteintech
22490-1-AP-150UL
10084-648EA 524.48 USD
10084-648
Anti-CEP290 Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: CEP290
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR