Anti-HNF1A Rabbit Polyclonal Antibody

Supplier: Proteintech
22426-1-AP-150UL
10088-362EA 402 USD
10088-362
Anti-HNF1A Rabbit Polyclonal Antibody
Antibodies
Western Blot:rat Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: HNF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR