Anti-Casein Kinase 2 beta Rabbit Polyclonal Antibody

Supplier: Proteintech
22418-1-AP-150UL
10082-066EA 402 USD
10082-066
Anti-Casein Kinase 2 beta Rabbit Polyclonal Antibody
Antibodies
Casein kinase II subunit beta is a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus. It participates in Wnt signaling, and plays a complex role in regulating the basal catalytic activity of the alpha subunit.

Western Blot: MCF-7 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR