Anti-AGXT Rabbit Polyclonal Antibody

Supplier: Proteintech
22394-1-AP-150UL
10082-616EA 402 USD
10082-616
Anti-AGXT Rabbit Polyclonal Antibody
Antibodies
IHC: Human Liver Tissue, 1:20-1:200; Western Blot: Mouse Liver Tissue, 1:500-1:5000

Type: Primary
Antigen: AGXT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR