Anti-IRF7 Rabbit Polyclonal Antibody

Supplier: Proteintech
22392-1-AP-150UL
10088-976EA 537.7 USD
10088-976
Anti-IRF7 Rabbit Polyclonal Antibody
Antibodies
IRF-7 (interferon regulatory factor 7) is a member of the interferon regulatory transcription factor (IRF) family. IRF-7 has been shown to play a role in the transcriptional activation of virus-inducible cellular proteins, including interferon beta chain proteins. Inducible expression of IRF-7 is largely restricted to lymphoid tissue. Four transcript variants encoding distinct isoforms (A,B,C and D) have been identified for this .The active IRF7A exists as a dimer form ~80 kDa. The MW 67-70 kDa has been reported in some papers .Various posttranslational modifications of IRF7 including phosphorylation, ubiquitination, sumoylation and acetylation are identified .This antibody is a rabbit polyclonal antibody raised against the C-terminal 349 amino acid residues of human IRF7 D.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: IRF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR