Anti-CXorf15 Rabbit Polyclonal Antibody

Supplier: Proteintech
22357-1-AP-150UL
10085-442EA 402 USD
10085-442
Anti-CXorf15 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Kidney Tissue, 1:500-1:5000

Type: Primary
Antigen: CXorf15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR