Anti-USP11 Rabbit Polyclonal Antibody

Supplier: Proteintech
22340-1-AP-150UL
10096-836EA 402 USD
10096-836
Anti-USP11 Rabbit Polyclonal Antibody
Antibodies
USP11(Ubiquitin carboxyl-terminal hydrolase 11) is also named as UHX1 and belongs to the peptidase C19 family. It is a deubiquitinating enzyme that controls many intracellular processes, including cell cycle progression, transcriptional activation, and signal transduction.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200

Type: Primary
Antigen: USP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR