Anti-ATP1B2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22338-1-AP-150UL
10083-270EA 402 USD
10083-270
Anti-ATP1B2 Rabbit Polyclonal Antibody
Antibodies
ATP1B2 is the β2 subunit of Na+/K+-ATPase which is an essential membrane-bound enzyme responsible for the transport of Na+ and K+ in most eukaryotic cells. ATP1B2 is also called the adhesion molecule on glia (AMOG) and it is highly expressed in normal glia. It is a heavily glycosylated protein that plays a role in cellular adhesion in the CNS. Recently differential expression of ATP1B2 has been found in some glioneuronal tumors (23887941, 19371356). This antibody recognizes the endogenous ATP1B2 protein in human brain. The bands between 45 kDa and 65kDa represent the glycosylated forms of ATP1B2 in different levels (8918259).

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: ATP1B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR