Anti-TCF4 Rabbit Polyclonal Antibody

Supplier: Proteintech
22337-1-AP-150UL
10095-726EA 402 USD
10095-726
Anti-TCF4 Rabbit Polyclonal Antibody
Antibodies
Transcription factor 4 (TCF4), also known as SEF2, binds to the immunoglobulin enchancer Mu-E5/KE5-motif. Involved in the initiation of neuronal differentiation. Activates transcription by binding to the E box (5'-CANNTG-3'). Binds to the E-box present in the somatostatin receptor 2 initiator element (SSTR2-INR) to activate transcription By similarity. Preferentially binds to either 5'-ACANNTGT-3' or 5'-CCANNTGG-3'.TCF4 has several splicing isoforms that are expressed differentially in tissues and during cancer progression(15547706, 12796377). This antibody reacts with 68kd and 72kd TCF4 proteins.

Western Blot: Mouse lung Tissue, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: TCF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR