Anti-RCC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22335-1-AP-150UL
10081-458EA 402 USD
10081-458
Anti-RCC1 Rabbit Polyclonal Antibody
Antibodies
CHC1, also named as RCC1, promotes the exchange of ran-bound gdp by gtp. It is involved in the regulation of onset of chromosome condensation in the S-phase. Phosphorylation of RCC1 on serines located in or near its nuclear localization signal activates RCC1 to generate RanGTP on mitotic chromosomes, which is required for spindle assembly and chromosome segregation. This antibody is a rabbit polyclonal antibody raised against full length RCC1 of human origin.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: RCC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR