Anti-PBX4 Rabbit Polyclonal Antibody

Supplier: Proteintech
22323-1-AP-150UL
10091-894EA 402 USD
10091-894
Anti-PBX4 Rabbit Polyclonal Antibody
Antibodies
Pre-B-cell leukemia homeobox 4 (PBX4), also named as Homeobox protein PBX4, is one of the pbx gene encode homeodomain containing transcriptional regulators that interact with other proteins to control embryogenesis and tumorigenesis. Pbx 4 expression is confined to the testis, especially to spermatocytes in the pachytene stage of the first meiotic prophase.PBX4 may form heterodimers with other homeobox genes (hoxb1b and meis3) to specify different developmental fates during vertebrate embryogenesis .The trimeric complex containing Pbx4, Meis3, and Hoxb1b was also formed.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: PBX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR