Anti-CC2D2A Rabbit Polyclonal Antibody

Supplier: Proteintech
22293-1-AP-150UL
10084-156EA 402 USD
10084-156
Anti-CC2D2A Rabbit Polyclonal Antibody
Antibodies
CC2D2A, also named as KIAA1345, may be involved in cilia formation. CC2D2A encodes a protein with similar overall structure to RPGRIP1L, including coiled-coil domains, a C2 domain, and an overlapping centrosomal protein-related domain. CC2D2A physically interacts with CEP290, and loss of Cc2d2a function in the zebrafish sentinel mutant results in abnormal body shape and pronephric (kidney) cysts that is strongly exacerbated by knockdown of Cep290 function. Defects in CC2D2A are the cause of Meckel syndrome type 6 (MKS6). Defects in CC2D2A are the cause of Joubert syndrome type 9 (JBTS9). The antibody is specific to CC2D2A.

Western Blot: HEK-293 Cells, 1:200-1:2000; IF: HTERT-RPE1 Cells, 1:20-1:200; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CC2D2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR