Anti-SAR1B Rabbit Polyclonal Antibody

Supplier: Proteintech
22292-1-AP-150UL
10081-482EA 402 USD
10081-482
Anti-SAR1B Rabbit Polyclonal Antibody
Antibodies
SAR1B, also named as SARA2, SARB and GTBPB, belongs to the small GTPase superfamily and SAR1 family. It is involved in transport from the endoplasmic reticulum to the Golgi apparatus. SAR1B is activated by the guanine nucleotide exchange factor PREB. It is involved in the selection of the protein cargo and the assembly of the COPII coat complex. The antibody is specific to SAR1B.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: SAR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR