Anti-ARF3 Rabbit Polyclonal Antibody

Supplier: Proteintech
22289-1-AP-150UL
10082-312EA 402 USD
10082-312
Anti-ARF3 Rabbit Polyclonal Antibody
Antibodies
ADP-ribosylation factors (ARFs) are members of the ARF family of GTP-binding proteins of the Ras superfamily, with 20kda protein size. ARFs bind and regulate GTP/GDP cycle by alternating between the active GTP-bound and inactive GDP-bound conformations. ARF family proteins are essential and ubiquitous in eukaryotes. Six highly conserved members of the family have been identified in mammalian cells. They function in vesicular traffic and actin remodelling and other bioprocesses in cells. ARF3 is 95% homolouous to ARF1. Its function was not fully understood. .The antibody is specific to ARF3. If has no cross reaction to other ARF members.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ARF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR