Anti-CLTCL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22283-1-AP-150UL
10084-886EA 402 USD
10084-886
Anti-CLTCL1 Rabbit Polyclonal Antibody
Antibodies
CLTCL1, also named as CLH22, CLTCL and CLTD, belongs to the clathrin heavy chain family. Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network. The antibody is specific to CLTCL1. It has no cross reaction to CLTC.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: CLTCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR