Anti-CALML6 Rabbit Polyclonal Antibody

Supplier: Proteintech
22274-1-AP-150UL
10083-988EA 402 USD
10083-988
Anti-CALML6 Rabbit Polyclonal Antibody
Antibodies
CALML6, also named as CAGLP, CALGP , belongs to the calmodulin family and Calglandulin subfamily. It contains 4 EF-hand domains. The antibody is specific to CALML6.

Western Blot: Human Heart Tissue, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: CALML6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR