Anti-RBM15B Rabbit Polyclonal Antibody

Supplier: Proteintech
22249-1-AP-150UL
10093-398EA 402 USD
10093-398
Anti-RBM15B Rabbit Polyclonal Antibody
Antibodies
RBM15B, one members of the SPEN (Split-end) family of proteins, have repressor function in several signaling pathways and may bind to RNA through interaction with spliceosome components. Present polyclonal anti-RBM15B antibody was produced by immunizing animals with C-terminus of RBM15B, which homolog with a short form of RBM15B(563aa, ~70kDa), thus this antibody can recognize full length of RBM15B(~100kDa) and splice-one(70kDa)

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: RBM15B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR