Anti-REST Rabbit Polyclonal Antibody

Supplier: Proteintech
22242-1-AP-150UL
10093-520EA 402 USD
10093-520
Anti-REST Rabbit Polyclonal Antibody
Antibodies
REST also named as NRSF or X2 box repressor is a 1097 amino acid protein, which contains 9 C2H2-type zinc fingers. REST as transcriptional repressor binds neuron-restrictive silencer element (NRSE) and represses neuronal gene transcription in non-neuronal cells. Repressor element 1 (RE1) silencing transcription factor (REST), is widely expressed during embryogenesis, and plays a strategic role in terminal neuronal differentiation. The molecular weight of REST is 121 kDa, but REST was detected 200kDa by western blot in our result, which is similar to the one article .

Western Blot:L02 Cells, 1:200-1:2000; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: REST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR