Anti-ADAM28 Rabbit Polyclonal Antibody

Supplier: Proteintech
22234-1-AP-150UL
10082-490EA 402 USD
10082-490
Anti-ADAM28 Rabbit Polyclonal Antibody
Antibodies
ADAM28 belongs to a family of cell surface and secreted glycoproteins that possess both proteolytic and adhesive properties. ADAM28 is predominantly expressed by carcinoma cells in the breast carcinoma tissues as protein bands of 42 kDa and 55/57 kDa, which correspond to the active forms of ADAM28s and ADAM28m, respectively.In human non-small cell lung carcinomas and breast carcinomas, ADAM28 is overexpressed predominantly by carcinoma cells, and the expression correlates with carcinoma cell proliferation and lymph node metastasis.

Western Blot: Mouse lung Tissue, 1:200-1:2000

Type: Primary
Antigen: ADAM28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR