Anti-FOXP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
22228-1-AP-150UL
10081-792EA 402 USD
10081-792
Anti-FOXP3 Rabbit Polyclonal Antibody
Antibodies
FOXP3, also named as IPEX, JM2 and Scurfin, is a transcription factor. POXP3 plays a critical role in the control of immune response. Defects in FOXP3 are the cause of immunodeficiency polyendocrinopathy, enteropathy, X-linked syndrome (IPEX) which also known as X-linked autoimmunity-immunodeficiency syndrome. This antibody is a rabbit polyclonal antibody raised against a peptide mapping within human FOXP3.

Western Blot: HeLa Cells, 1:500-1:5000; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR