Anti-Rabenosyn 5 Rabbit Polyclonal Antibody

Supplier: Proteintech
22218-1-AP-150UL
10093-238EA 402 USD
10093-238
Anti-Rabenosyn 5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: Rabenosyn 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR