Anti-ACP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22214-1-AP-150UL
10082-446EA 402 USD
10082-446
Anti-ACP1 Rabbit Polyclonal Antibody
Antibodies
Human red cell acid phosphatase (ACP1) is a polymorphic enzyme closely related to cytosolic low molecular weight acid phosphatases, a protein family broadly conserved among eukaryotes. It catalyses the transfer of phosphate from phosphate ester substrates to suitable acceptor alcohols such as methanol and glycerol. This protein has 3 isoforms produced by alternative splicing.

  • Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ACP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR