Anti-TMF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22212-1-AP-150UL
10095-706EA 402 USD
10095-706
Anti-TMF1 Rabbit Polyclonal Antibody
Antibodies
TMF1, also named as TMF and ARA160, is a potential coactivator of the androgen receptor. It mediates STAT3 degradation. The MW of TMF1 is about 160 kDa.

Western Blot: Mouse Brain Tissue, 1:1000-1:10000; IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: TMF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR