Anti-ERC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22211-1-AP-150UL
10086-606EA 402 USD
10086-606
Anti-ERC1 Rabbit Polyclonal Antibody
Antibodies
ERC is an acronym based on previous separate namings of members of this protein family, ELKS, Rab6IP2 and CAST . They are known to bind RIMs, the active zone proteins that regulate neurotransmitter release. ERC1 (ELKS) is an essential regulatory subunit of the IKK complex and likely functions by recruiting IκBα to the complex . Multiple isoforms of ERC1 mRNA are generated by alternative splicing . There are two known splice variants of ERC1 protein that differ at their C termini, ERC1a and ERC1b. ERC1a is ubiquitously expressed, whereas ERC1b is brain-specific .

Western Blot: HeLa Cells, 1:500-1:5000; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: ERC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR