Anti-CK7 Rabbit Polyclonal Antibody

Supplier: Proteintech
22208-1-AP-150UL
10081-540EA 402 USD
10081-540
Anti-CK7 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:1000-1:10000; IF: HeLa Cells, 1:10-1:100; FC: HeLa Cells, N/A; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR