Anti-ZAP70 Rabbit Polyclonal Antibody

Supplier: Proteintech
22207-1-AP-150UL
10081-348EA 402 USD
10081-348
Anti-ZAP70 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Lymphoma Tissue, 1:20-1:200; Western Blot: RAW 264.7 Cells, 1:500-1:5000

Type: Primary
Antigen: ZAP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR