Anti-CHRNB4 Rabbit Polyclonal Antibody

Supplier: Proteintech
22192-1-AP-150UL
10084-750EA 402 USD
10084-750
Anti-CHRNB4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:1000-1:10000

Type: Primary
Antigen: CHRNB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR