Anti-CCZ1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22159-1-AP-150UL
10084-260EA 402 USD
10084-260
Anti-CCZ1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:NIH/3T3 Cells, 1:500-1:5000; IP:NIH/3T3 Cells, 1:200-1:2000

Type: Primary
Antigen: CCZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR