Anti-ADAM5 Rabbit Polyclonal Antibody

Supplier: Proteintech
22147-1-AP-150UL
10082-492EA 402 USD
10082-492
Anti-ADAM5 Rabbit Polyclonal Antibody
Antibodies
ADAM5(putative disintegrin and metalloproteinase domain-containing protein 5) is also named as ADAM5P, TMDC2(tMDCII). The tMDC II, abundantly expressed, sperm surface MDC proteins identified in rodent and macaque systems, is also nonfunctional in the human. It has 7 isoforms produced by alternative splicing. It is expressed in testis highly by northern blot. This antibody is specific to ADAM5.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: ADAM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR