Anti-RASAL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22140-1-AP-150UL
10093-344EA 402 USD
10093-344
Anti-RASAL2 Rabbit Polyclonal Antibody
Antibodies
RASAL2 (Ras protein activator like 2), also known as nGAP, is a Ras GTPase-activating protein, containing a GRD domain and a coiled-coil structure at the C terminus. It is able to complement the defective RasGAP function in a yeast system. RASAL2 is highly homologous to Ras-specific GTPase activating proteins (RasGAPs). Two transcript variants exist for RASAL2 due to alternative splicing events.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: RASAL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR