Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: Proteintech
22104-1-AP-150UL
10081-824EA 537.7 USD
10081-824
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
Glycogen synthase kinase-3 (GSK3) is a proline-directed serine-threonine kinase that was initially identified as a phosphorylating and inactivating glycogen synthase .GSK3B is involved in energy metabolism, neuronal cell development, and body pattern formation.In skeletal muscle, it contributes to insulin regulation of glycogen synthesis by phosphorylating and inhibiting GYS1 activity and hence glycogen synthesis. Researches showed that the crystal structure of human GSK3B, expressed in insect cells, at 2.8-angstrom resolution .This antibody recognize the C-terminal of GSK3B.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR