Anti-CCR10 Rabbit Polyclonal Antibody

Supplier: Proteintech
22071-1-AP-150UL
10084-230EA 402 USD
10084-230
Anti-CCR10 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human spleen Tissue, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: CCR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR