Anti-CDK8 Rabbit Polyclonal Antibody

Supplier: Proteintech
22067-1-AP-150UL
10084-532EA 453 USD
10084-532
Anti-CDK8 Rabbit Polyclonal Antibody
Antibodies
Western Blot:NIH/3T3 Cells, 1:500-1:5000; IHC: Human Stomach cancer, 1:20-1:200

Type: Primary
Antigen: CDK8
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR