Anti-CDH20 Rabbit Polyclonal Antibody

Supplier: Proteintech
22049-1-AP-150UL
10083-954EA 402 USD
10083-954
Anti-CDH20 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: CDH20
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR