Anti-B3GALNT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22048-1-AP-150UL
10083-440EA 402 USD
10083-440
Anti-B3GALNT1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Placenta Tissue, 1:500-1:5000; IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: B3GALNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR