Anti-NPHP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
22026-1-AP-150UL
10091-248EA 402 USD
10091-248
Anti-NPHP3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Testis Tissue, 1:200-1:2000; IF: HTERT-RPE1 Cells, 1:20-1:200; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: NPHP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR