Anti-ARPC5L Rabbit Polyclonal Antibody

Supplier: Proteintech
22025-1-AP-150UL
10083-150EA 402 USD
10083-150
Anti-ARPC5L Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: ARPC5L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR