Anti-CDH2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22018-1-AP-150UL
10081-656EA 537.7 USD
10081-656
Anti-CDH2 Rabbit Polyclonal Antibody
Antibodies
Cadherins are a family of transmembrane glycoproteins that mediate calcium-dependent cell-cell adhesion and play an important role in the maintenance of normal tissue architecture. N-cadherin (neural cadherin), also known as CDH2 (cadherin 2), is a classical member of the cadherin superfamily which also include E-, P-, R-, and B-cadherins. Expression of N-cadherin has been reported on various cell types including neurons, endothelial cells and cardiac myocytes . N-cadherin has functions in early brain morphogenesis, synaptogenesis and synaptic plasticity .

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IP: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; FC:SH-SY5Y Cells, N/A

Type: Primary
Antigen: CDH2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR