Anti-VAT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22016-1-AP-150UL
10096-934EA 402 USD
10096-934
Anti-VAT1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:1000-1:10000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: VAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR