Anti-MYCT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22004-1-AP-150UL
10090-684EA 402 USD
10090-684
Anti-MYCT1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse spleen Tissue, 1:500-1:5000

Type: Primary
Antigen: MYCT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR