Anti-STRA6 Rabbit Polyclonal Antibody

Supplier: Proteintech
22001-1-AP-150UL
10095-346EA 402 USD
10095-346
Anti-STRA6 Rabbit Polyclonal Antibody
Antibodies
Specific

Type: Primary
Antigen: STRA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR