Anti-ACC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21923-1-AP-150UL
10082-416EA 537.7 USD
10082-416
Anti-ACC1 Rabbit Polyclonal Antibody
Antibodies
ACACA(Acetyl-CoA carboxylase 1, ACC), also named as ACAC, ACC1 and ACCA, belongs to the biotin containing enzyme family. It catalyzes the synthesis of malonyl-CoA, which is an intermediate substrate playing a pivotal role in the regulation of fatty acid metabolism and energy production. ACACA is involved in the biosynthesis of fatty acids, and malonyl-CoA produced is used as a building block to extend the chain length of fatty acids by fatty acid synthase (FAS). It has 4 isoforms produced by alternative promoter usage with the molecular weight between 260 kDa and 270 kDa.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: ACC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR