Anti-OAS3 Rabbit Polyclonal Antibody

Supplier: Proteintech
21915-1-AP-150UL
10091-518EA 402 USD
10091-518
Anti-OAS3 Rabbit Polyclonal Antibody
Antibodies
The 2-prime,5-prime oligoadenylate synthetases (OASs), such as OAS3, are interferon-induced proteins characterized by their capacity to catalyze the synthesis of 2-prime,5-prime oligomers of adenosine (2-5As). It is also named as p100 OAS and belongs to the 2-5A synthase family. OAS3 is involved in inhibition of cellular protein synthesis and in viral infection resistance.

Western Blot:A375 Cells, 1:200-1:2000; IHC: Human skin cancer Tissue, 1:20-1:200

Type: Primary
Antigen: OAS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR