Anti-DUS4L Rabbit Polyclonal Antibody

Supplier: Proteintech
21870-1-AP-150UL
10086-168EA 402 USD
10086-168
Anti-DUS4L Rabbit Polyclonal Antibody
Antibodies
tRNA-dihydrouridine synthase 4-like belongs to the dus family.It catalyzes the synthesis of dihydrouridine and a modified base found in the D-loop of most tRNAs.Researches showed that DUS4L was alsophosphorylated in vitro by a Ca2+ calmodulin-activated kinase. It has 2 isoforms produced by alternative splicing .One, at 35 kDa, comigrated with a second cAMP-stimulated RIS-ROS phosphoprotein,DUS4L,which was also detected in the cytoskeleton.This antibody recognize the C-terminal of DUS4L.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000

Type: Primary
Antigen: DUS4L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR