Anti-SAFB Rabbit Polyclonal Antibody

Supplier: Proteintech
21857-1-AP-150UL
10094-114EA 402 USD
10094-114
Anti-SAFB Rabbit Polyclonal Antibody
Antibodies
Heterogeneous nuclear ribonucleoproteins (hnRNPs) constitute a set of polypeptides that contribute to pre-mRNA processing and transport. hnRNPs also bind heterogeneous nuclear RNA (hnRNA), the transcripts produced by RNA polymerase II. SAFB (scaffold attachment factor B) is a nuclear matrix-associated protein that binds to matrix- or scaffold-associating regions (MARs or SARs) on DNA and interacts with RNA polymerase II and serine-/arginine-rich RNA processing factors (SR proteins). SAFB, also designated HAP (hnRNP A1 associated protein) and HET (HSP 27-ERE-TATA-binding protein) is a proven hnRNP protein that has a speckled distribution in the nucleus and, in response to stress agents such as heat shock, is recruited to a few, large nuclear granules, called perichromatin granules. SAFB also binds to the estrogen receptor (ER) and is expressed in several breast cancer cell lines at varying levels. Subsequently, SAFB may play a role in breast cancer by mediating cellular proliferation and division. This antibody specifically reacts whith the isoform 1 of human SAFB protein.

Western Blot: MCF-7 Cells, 1:500-1:5000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: SAFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR