Anti-KIDINS220 Rabbit Polyclonal Antibody

Supplier: Proteintech
21856-1-AP-150UL
10089-212EA 402 USD
10089-212
Anti-KIDINS220 Rabbit Polyclonal Antibody
Antibodies
Kidins220 (Kinase D-interacting substrate 220 kD) is a crucial mediator of signal transduction in neural tissues. Also it named ARMS(Ankyrin Repeat-Rich Membrane Spanning) due to the presence of ankyrin repeats and transmembrane domains together with other binding motifs. It is a multidomain transmembrane protein that not only transduces signaling mediated by the receptor tyrosine kinase (RTK), but also regulates the stability of cytoskeletal network through a number of cytoskeleton-associated proteins, including Septin5, MAPs , and stathmin family members.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: KIDINS220
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR