Anti-GLUT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
21829-1-AP-150UL
10094-558EA 537.7 USD
10094-558
Anti-GLUT1 Rabbit Polyclonal Antibody
Antibodies
SLC2A1, also known as GLUT1, is an ubiquitously expressed glucose transporter and responsible for the basal level of glucose uptake in most cell types. Human erythrocytes express the highest level of the SLC2A1. Defects in SLC2A1 are the cause of GLUT1 deficiency syndrome type 1 and type 2. High expression of SLC2A1 has been reported to be a reliable immunohistochemical marker for juvenile hemangiomas.

Western Blot:Y79 Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HeLa Cells, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: GLUT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR