Anti-CEP152 Rabbit Polyclonal Antibody

Supplier: Proteintech
21815-1-AP-150UL
10084-586EA 402 USD
10084-586
Anti-CEP152 Rabbit Polyclonal Antibody
Antibodies
CEP152, also named as KIAA0912, is a regulator of genomic integrity and cellular response to DNA damage acting through ATR-mediated checkpoint signaling. It is necessary for centrosome duplication. CEP152 functions as a molecular scaffold facilitating the interaction of PLK4 and CENPJ, two molecules involved in centriole formation. CEP152, ATR and CENPJ control cellular responses to DNA damage. CEP152 has some isoforms woth MW 189 kDa, 152 kDa, 110 kDa and 66 kDa.

IP: Mouse Brain Tissue, 1:200-1:2000; Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: CEP152
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR