Anti-LDHA Rabbit Polyclonal Antibody

Supplier: Proteintech
21799-1-AP-150UL
10089-568EA 402 USD
10089-568
Anti-LDHA Rabbit Polyclonal Antibody
Antibodies
LDHA, also named as PIG19, belongs to the LDH/MDH superfamily and LDH family. Defects in LDHA are the cause of glycogen storage disease type 11 (GSD11).It has 5 isoforms and can form a homotetramer .

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR